missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzyl alcohol, MedChemExpress

Product Code. 16429426
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16429426 Supplier Medchem Express Supplier No. HYY0892

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 4-Hydroxybenzyl alcohol is a phenolic compound widely distributed in various kinds of plants. Anti-inflammatory, anti-oxidant, anti-nociceptive activity. Neuroprotective effect. Inhibitor of tumor angiogenesis and growth.

4-Hydroxybenzyl alcohol is a phenolic compound widely distributed in various kinds of plants. Anti-inflammatory, anti-oxidant, anti-nociceptive activity. Neuroprotective effect. Inhibitor of tumor angiogenesis and growth.

Specifications

Chemical Name or Material 4-Hydroxybenzyl alcohol
CAS 623-05-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H8O2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (805.54 mM; Need ultrasonic)
SMILES OCC1=CC=C(O)C=C1
Molecular Weight (g/mol) 124.14
Formula Weight 124.14
Percent Purity 99.34%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.