missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzyl alcohol, 99%

Product Code. 11454494
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454494 250 g 250g
11444494 50 g 50g
11374709 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454494 Supplier Thermo Scientific Alfa Aesar Supplier No. A13132.30
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 623-05-2 | C7H8O2 | 124.139 g/mol

4-Hydroxybenzyl alcohol is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzyl alcohol is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (6.7 mg/ml at 20°C), dioxane (100 mg/ml), 1N NaOH (50 mg/ml), DMSO, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acid chlorides, acid anhydrides, bases, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-05-2
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00004658
InChI Key BVJSUAQZOZWCKN-UHFFFAOYSA-N
Synonym 4-hydroxybenzyl alcohol, 4-hydroxymethyl phenol, p-hydroxybenzyl alcohol, p-methylolphenol, 4-methylolphenol, 4-hydroxybenzenemethanol, benzenemethanol, 4-hydroxy, p-hydroxymethyl phenol, gastrodigenin, 4-hydroxybenzylalcohol
PubChem CID 125
ChEBI CHEBI:67410
IUPAC Name 4-(hydroxymethyl)phenol
SMILES C1=CC(=CC=C1CO)O

Specifications

Melting Point 114°C to 122°C
Boiling Point 251°C to 253°C
Quantity 250 g
Beilstein 1858967
Sensitivity Air and light sensitive
Solubility Information Soluble in water (6.7mg/ml at 20°C),dioxane (100mg/ml),1N NaOH (50mg/ml),DMSO,and methanol.
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 4-Hydroxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.