missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzyl alcohol, 97%

Product Code. 10336900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336900 100 g Glass bottle 100g
10784431 25 g Glass bottle 25g
11934671 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10336900 Supplier Thermo Scientific Acros Supplier No. 121031000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-05-2 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-05-2
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
InChI Key BVJSUAQZOZWCKN-UHFFFAOYSA-N
Synonym 4-hydroxybenzyl alcohol, 4-hydroxymethyl phenol, p-hydroxybenzyl alcohol, p-methylolphenol, 4-methylolphenol, 4-hydroxybenzenemethanol, benzenemethanol, 4-hydroxy, p-hydroxymethyl phenol, gastrodigenin, 4-hydroxybenzylalcohol
PubChem CID 125
ChEBI CHEBI:67410
IUPAC Name 4-(hydroxymethyl)phenol
SMILES C1=CC(=CC=C1CO)O

Specifications

Melting Point 110.0°C to 112.0°C
Color Beige to Pink
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H4CH2OH
Quantity 100 g
Beilstein 06,897
Solubility Information Solubility in water: soluble
Formula Weight 124.14
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzyl alcohol
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 4-Hydroxybenzyl alcohol
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.