missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzotrifluoride, LoGiCal

Product Code. 30458159
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30458159 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30458159

Brand: LoGiCal LGCFOR0256.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxybenzotrifluoride
CAS 402-45-9
Analyte or Component Names 4-Hydroxybenzotrifluoride
Shipping Condition Room Temperature
Molecular Formula C7 H5 F3 O
Certifications/Compliance ISO 9001
Synonym 4-(Trifluoromethyl)phenol (ACI), p-Cresol, α,α,α-trifluoro- (7CI, 8CI), NSC 88303, p-Hydroxybenzotrifluoride, p-Trifluoromethylphenol, α,α,α-Trifluoro-4-cresol, α,α,α-Trifluoro-p-cresol
InChI Formula InChI=1S/C7H5F3O/c8-7(9,10)5-1-3-6(11)4-2-5/h1-4,11H
Solution Type Neat
SMILES Oc1ccc(cc1)C(F)(F)F
IUPAC Name 4-(trifluoromethyl)phenol
Industry Type Clinical Testing
Molecular Weight (g/mol) 162.11
Quantity 10 mg
Formula Weight 162.0292 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.