missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzophenone, TRC

Product Code. 30395061
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395061 100 g
30401517 25 g
30398578 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395061 Supplier Toronto Research Chemicals Supplier No. TRCH829305100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1137-42-4
Molecular Formula C13 H10 O2
Molecular Weight (g/mol) 198.2173
Synonym Benzophenone, 4-hydroxy- (6CI,7CI,8CI), Benzophenone, p-hydroxy- (3CI), (4-Hydroxyphenyl)phenylmethanone, 1-(4-Hydroxyphenyl)-1-phenylmethanone, 4-Benzoylphenol, 4-Hydroxybenzophenone, 4-Hydroxyphenyl phenyl ketone, 4'-Hydroxybenzophenone, N-(4-Hydroxylphenyl)-N'-phenylurea, NSC 1887, p-Benzoylphenol, p-Hydroxybenzophenone
IUPAC Name (4-hydroxyphenyl)-phenylmethanone
SMILES Oc1ccc(cc1)C(=O)c2ccccc2

Specifications

Quantity 100 g
InChI Formula InChI=1S/C13H10O2/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9,14H
Formula Weight 198.0681
Percent Purity >95%
Chemical Name or Material 4-Hydroxybenzophenone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.