missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzoic Acid, Mikromol™

Product Code. 17961696
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17961696 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17961696 Supplier Mikromol Supplier No. MM0045.010250
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Hydroxybenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxybenzoic Acid
CAS 99-96-7
Analyte or Component Names 4-Hydroxybenzoic Acid
Shipping Condition Room Temperature
Molecular Formula C7 H6 O3
Certifications/Compliance ISO 17034
Synonym 4-Hydroxybenzoic Acid, Ethyl Parahydroxybenzoate Imp. A (EP), Butyl Parahydroxybenzoate Imp. A (EP), Propyl Parahydroxybenzoate Imp. A (EP), Methyl Parahydroxybenzoate Imp. A (EP), Acetylsalicylic Acid Imp. A (EP), Salicylic Acid Imp. A (EP), 4-Carboxyphenol, 4-Hydroxybenzoic acid, NSC 4961, Paraben-acid, p-Carboxyphenol, p-Hydroxybenzoic acid, p-Salicylic acid, Salicylic Acid USP Related Compound A, Salicylic Acid USP RC A, SKF-29685, CCI 23875, Ph Eur Salicylic Acid Impurity A, GW 466608X, GW 470921X, GW 403126X, GW 357662X, GW 444710X, Salicylic Acid Related Compound A, Acetylsalicylic Acid Impurity A, Butyl Parahydroxybenzoate Impurity A, Ethyl Parahydroxybenzoate Impurity A, Methyl Parahydroxybenzoate Impurity A, Propyl Parahydroxybenzoate Impurity A, Salicylic Acid Impurity A
InChI Formula InChI=1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)
SMILES OC(=O)c1ccc(O)cc1
IUPAC Name 4-hydroxybenzoic acid
Molecular Weight (g/mol) 138.12
Quantity 250 mg
Formula Weight 138.0317 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.