missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzoic acid-methyl ester, Dr. Ehrenstorfer

Product Code. 30328321
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328321 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328321 Supplier Dr. Ehrenstorfer Supplier No. DREC14229000100MG
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Parahydroxybenzoate
CAS 99-76-3
Analyte or Component Names Methyl Parahydroxybenzoate
Shipping Condition +20°C
Molecular Formula C8 H8 O3
Certifications/Compliance ISO 17034
Synonym Methyl 4-Hydroxybenzoate, Methyl Parahydroxybenzoate, Nifuroxazide Imp. B (EP), Ethyl Parahydroxybenzoate Imp. B (EP), Butyl Parahydroxybenzoate Imp. B (EP), Propyl Parahydroxybenzoate Imp. B (EP), Methyl Paraben, Methylparaben, SKF-4382, Methyl Parahydroxybenzoate, Nipagin, Methyl Paraben, 4-Hydroxybenzoic acid-methyl ester, Butyl Parahydroxybenzoate Impurity B, Ethyl Parahydroxybenzoate Impurity B, Methyl Salicylate Impurity C, Nifuroxazide Impurity B, Propyl Parahydroxybenzoate Impurity B
InChI Formula InChI=1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3
SMILES COC(=O)c1ccc(O)cc1
IUPAC Name methyl 4-hydroxybenzoate
Molecular Weight (g/mol) 152.15
Quantity 100 mg
Formula Weight 152.0473 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.