missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzoic acid-methyl ester 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30316996
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316996 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316996 Supplier Dr. Ehrenstorfer Supplier No. DREA14229000AL1000
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Parahydroxybenzoate
CAS 99-76-3
Analyte or Component Names Methyl Parahydroxybenzoate
Shipping Condition Room Temperature
Molecular Formula C8 H8 O3
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Methyl 4-Hydroxybenzoate, Methyl Parahydroxybenzoate, Nifuroxazide Imp. B (EP), Ethyl Parahydroxybenzoate Imp. B (EP), Butyl Parahydroxybenzoate Imp. B (EP), Propyl Parahydroxybenzoate Imp. B (EP), Methyl Paraben, Methylparaben, SKF-4382, Methyl Parahydroxybenzoate, Nipagin, Methyl Paraben, 4-Hydroxybenzoic acid-methyl ester, Butyl Parahydroxybenzoate Impurity B, Ethyl Parahydroxybenzoate Impurity B, Methyl Salicylate Impurity C, Nifuroxazide Impurity B, Propyl Parahydroxybenzoate Impurity B
InChI Formula InChI=1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3
SMILES COC(=O)c1ccc(O)cc1
IUPAC Name methyl 4-hydroxybenzoate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 152.15
Quantity 1 mL
Formula Weight 152.0473 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.