missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzoic Acid Hydrazide, 98%

Product Code. 10318793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10318793 500 g Plastic bottle 500g
10122250 25 g Plastic bottle 25g
10487690 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10318793 Supplier Thermo Scientific Acros Supplier No. 121005000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5351-23-5 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5351-23-5
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007605
InChI Key ZMZGIVVRBMFZSG-UHFFFAOYSA-N
Synonym 4-hydroxybenzhydrazide, 4-hydroxybenzoic acid hydrazide, 4-hydroxybenzoic hydrazide, p-hydroxybenzhydrazide, p-hydroxybenzoic acid hydrazide, 4-hydroxybenzoylhydrazine, benzoic acid, 4-hydroxy-, hydrazide, p-hydroxybenzoic hydrazide, p-hydroxybenzoyl hydrazine, 4-hydroxy-benzoic acid hydrazide
PubChem CID 1742
IUPAC Name 4-hydroxybenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 260.0°C to 264.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula HOC6H4CONHNH2
Quantity 500 g
Beilstein 10, 174
Solubility Information Solubility in water: 26.5g/L (25°C). Other solubilities: soluble in methanol,ethanol
Formula Weight 152.15
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Hydroxybenzoic acid hydrazide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.