missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzoic acid, 99%

Product Code. 11414694
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11414694

Brand: Thermo Scientific Alfa Aesar A13700.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 99-96-7 | C7H6O3 | 138.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Hydroxybenzoic acid is used as an intermediate for dyes, antiseptics and active pharmaceutical ingredients. It is also used as a food preservative, corrosion inhibitor, anti-oxidant and an emulsifier. It reacts with 6-hydroxynaphthalene-2-carboxylic acid to prepare aromatic polyester called Vectran fiber. It acts as a precursor to prepare p-hydroxyphenylbenzoate, p-acetoxybenzoyl chloride and 4,4′-dihydroxybenzophenone. Further, it is used in paint additives, coating additives, processing aids, electrical and electronic products. In addition to this, it acts as a preservative in cosmetics and some ophthalmic solutions.

Solubility
Soluble in water, alcohol, ethanol, ether, n-butanol and acetone. Slightly soluble in chloroform. Insoluble in carbon disulfide.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-96-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002547
InChI Key FJKROLUGYXJWQN-UHFFFAOYSA-N
Synonym p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german
PubChem CID 135
ChEBI CHEBI:30763
IUPAC Name 4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 213°C to 218°C
Flash Point 199°C (390°F)
Assay Percent Range 99%
Quantity 1000 g
Beilstein 970950
Merck Index 14,4812
Solubility Information Soluble in water,alcohol,ethanol,ether,n-butanol and acetone. Slightly soluble in chloroform. Insoluble in carbon disulfide.
Formula Weight 138.12
Percent Purity 99%
Chemical Name or Material 4-Hydroxybenzoic acid
compliance-icons compliance-icons
Product Identifier
  • 4-Hydroxybenzoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : p-Hydroxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.