missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzenesulfonyl Chloride, TRC

Product Code. 30397040
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397040 1 g 1g
30401500 250 mg 250mg
30391384 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397040 Supplier Toronto Research Chemicals Supplier No. TRCH9999631G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4025-67-6
Molecular Formula C6H5O3SCl
Molecular Weight (g/mol) 192.62
IUPAC Name 4-hydroxybenzenesulfonyl chloride
SMILES Oc1ccc(cc1)S(=O)(=O)Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H5ClO3S/c7-11(9,10)6-3-1-5(8)2-4-6/h1-4,8H
Formula Weight 191.9648
Chemical Name or Material 4-Hydroxybenzenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.