missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzenesulfonic Acid Sodium Salt, TRC

Product Code. 30389610
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30389610 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389610 Supplier Toronto Research Chemicals Supplier No. TRCH80781010G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxybenzenesulfonic Acid Sodium Salt
CAS 825-90-1
Molecular Formula C6H5NaO4S
Quantity 10 g
Synonym Sodium 4-Hydroxybenzenesulfonate, Sodium p-Hydroxybenzenesulfonate, Sodium p-Hydroxyphenylsulfonate, Sodium p-Phenolsulfonate, p-Hydroxybenzenesulfonic Acid Monosodium Salt, p-Hydroxybenzenesulfonic Acid Sodium Salt, p-Phenolsulfonic Acid Sodium Salt
InChI Formula InChI=1S/C6H6O4S.Na/c7-5-1-3-6(4-2-5)11(8,9)10;/h1-4,7H,(H,8,9,10);/q;+1/p-1
SMILES [Na+].Oc1ccc(cc1)S(=O)(=O)[O-]
IUPAC Name sodium;4-hydroxybenzenesulfonate
Molecular Weight (g/mol) 196.16
Formula Weight 195.9806
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.