missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzenesulfonic Acid Ammonium Salt, TRC

Product Code. 30399983
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399983 1 g 1g
30400110 50 g -
30390184 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399983 Supplier Toronto Research Chemicals Supplier No. TRCH8078151G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 856777-15-6
Molecular Formula C6H9NO4S
Molecular Weight (g/mol) 191.2
Synonym p-Hydroxy-benzenesulfonic Acid Ammonium Salt
IUPAC Name ammonium 4-hydroxybenzenesulfonate
SMILES O=S(C1=CC=C(O)C=C1)([O-])=O.[NH4+]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H6O4S.H3N/c7-5-1-3-6(4-2-5)11(8,9)10;/h1-4,7H,(H,8,9,10);1H3
Formula Weight 191.03
Chemical Name or Material 4-Hydroxybenzenesulfonic Acid Ammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.