missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzeneboronic Acid, TRC

Product Code. 30403494
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403494 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403494 Supplier Toronto Research Chemicals Supplier No. TRCH9495552.5G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxybenzeneboronic Acid
CAS 71597-85-8
Molecular Formula C6H7BO3
Quantity 2.5 g
Synonym B-(4-hydroxyphenyl)-boronic acid, p-Hydroxy-benzeneboronic Acid, (4-Hydroxyphenyl)-boronic Acid, (4-Hydroxyphenyl)boronic Acid, (p-Hydroxyphenyl)boronic Acid, 4-Hydroxybenzeneboronic Acid
InChI Formula InChI=1S/C6H7BO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8-10H
SMILES OB(O)c1ccc(O)cc1
IUPAC Name (4-hydroxyphenyl)boronic acid
Molecular Weight (g/mol) 137.93
Formula Weight 138.0488
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.