Learn More
4-Hydroxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
Description
4-Hydroxybenzeneboronic acid is used for Suzuki-Miyaura coupling and Stille coupling and potential for introducing different alkyl groups, palladium-catalyzed aminocarbonylation and cross-coupling reactions, bio-supported palladium nanoparticles as phosphine-free catalyst for Suzuki reaction in water and Cu2O-catalyzed aerobic oxidative cross-coupling of tetrazoles. It is also used in preparation of rod-like dendronized polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Hydroxybenzeneboronic acid is used for Suzuki-Miyaura coupling and Stille coupling and potential for introducing different alkyl groups, palladium-catalyzed aminocarbonylation and cross-coupling reactions, bio-supported palladium nanoparticles as phosphine-free catalyst for Suzuki reaction in water and Cu2O-catalyzed aerobic oxidative cross-coupling of tetrazoles. It is also used in preparation of rod-like dendronized polymers.
Solubility
Soluble in water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Melting Point | ∽285°C (decomposition) |
Quantity | 1 g |
Beilstein | 2963491 |
Solubility Information | Soluble in water |
Formula Weight | 137.93 |
Percent Purity | 97% |
Chemical Name or Material | 4-Hydroxybenzeneboronic acid |
RUO – Research Use Only