missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzaldehyde, 98%

Product Code. 11410104
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410104 500 g 500g
11304719 25 g 25g
11400104 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410104 Supplier Thermo Scientific Alfa Aesar Supplier No. A13580.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-08-0 | C7H6O2 | 122.123 g/mol

4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzaldehyde is used as a chemical that maintains bactericidal activity when tested against certain bacteria strains.

Solubility
Soluble in water(13.8g/L), ether, alcohol, and acetone.

Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 123-08-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00006939
InChI Key RGHHSNMVTDWUBI-UHFFFAOYSA-N
Synonym p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde
PubChem CID 126
ChEBI CHEBI:17597
IUPAC Name 4-hydroxybenzaldehyde
SMILES C1=CC(=CC=C1C=O)O

Spezifikation

Melting Point 115°C to 119°C
Density 1.129
Boiling Point 191°C (50 mmHg)
Flash Point 174°C (345°F)
Quantity 500 g
Beilstein 471352
Sensitivity Air Sensitive
Merck Index 14,4811
Solubility Information Soluble in water(13.8g/L),ether,alcohol,and acetone.
Formula Weight 122.12
Percent Purity 98%
Chemical Name or Material 4-Hydroxybenzaldehyde
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.