missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyazobenzene-4-carboxylic acid hydrate, TRC

Product Code. 30389965
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389965 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389965 Supplier Toronto Research Chemicals Supplier No. TRCH8052031G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Hydroxyazobenzene-4-carboxylic acid hydrate
CAS 1899039-20-3
Molecular Formula C13H12N2O4
Quantity 1 g
InChI Formula InChI=1S/C13H10N2O3.H2O/c16-12-7-5-11(6-8-12)15-14-10-3-1-9(2-4-10)13(17)18;/h1-8,16H,(H,17,18);1H2
SMILES O=C(O)C1=CC=C(N=NC2=CC=C(O)C=C2)C=C1.[H]O[H]
IUPAC Name 4-((4-hydroxyphenyl)diazenyl)benzoic acid hydrate
Molecular Weight (g/mol) 260.25
Formula Weight 260.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.