missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyacetophenone, TRC

Product Code. 30408152
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30408152 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30408152

Brand: Toronto Research Chemicals TRCH7399805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxyacetophenone
CAS 99-93-4
Purity Grade Notes HPLC
Molecular Formula C8 H8 O2
Quantity 5 g
Synonym 1-(4-Hydroxyphenyl)ethanone, Paracetamol Imp. E (EP), 4-Hydroxyacetophenone
InChI Formula InChI=1S/C8H8O2/c1-6(9)7-2-4-8(10)5-3-7/h2-5,10H,1H3
SMILES CC(=O)c1ccc(O)cc1
IUPAC Name 1-(4-hydroxyphenyl)ethanone
Molecular Weight (g/mol) 136.15
Formula Weight 136.0524
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.