missing translation for 'onlineSavingsMsg'
Learn More

4’-Hydroxy Trazodone-d6 Benzyl Ether, TRC

Product Code. 30399615
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399615 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30399615 Supplier Toronto Research Chemicals Supplier No. TRCH9700321MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Hydroxy Trazodone-d6 Benzyl Ether
CAS 1794752-37-6
Molecular Formula C26 2H6 H22 Cl N5 O2
Quantity 1 mg
Synonym 2-[3-[4-(3-Chloro-4-benzyloxyphenyl)-1-piperazinyl]propyl-d6]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one, 2-[3-[4-(m-Chloro-p-benzyloxyphenyl)-1-piperazinyl]propyl-d6]-s-triazolo[4.3-a]pyridin-3(2H)-one, 4-benzyloxytrazodone-d6
InChI Formula InChI=1S/C26H28ClN5O2/c27-23-19-22(10-11-24(23)34-20-21-7-2-1-3-8-21)30-17-15-29(16-18-30)12-6-14-32-26(33)31-13-5-4-9-25(31)28-32/h1-5,7-11,13,19H,6,12,14-18,20H2/i6D2,12D2,14D2
SMILES [2H]C([2H])(N1CCN(CC1)c2ccc(OCc3ccccc3)c(Cl)c2)C([2H])([2H])C([2H])([2H])N4N=C5C=CC=CN5C4=O
IUPAC Name 2-[3-[4-(3-chloro-4-phenylmethoxyphenyl)piperazin-1-yl]-1,1,2,2,3,3-hexadeuteriopropyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one
Molecular Weight (g/mol) 484.023
Formula Weight 483.231
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.