missing translation for 'onlineSavingsMsg'
Learn More

4’-Hydroxy Toremifene-d6, TRC

Product Code. 30396142
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396142 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30396142 Supplier Toronto Research Chemicals Supplier No. TRCH9545121MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Hydroxy Toremifene-d6
CAS 1795138-08-7
Molecular Formula C26 2H6 H22 Cl N O2
Quantity 1 mg
Synonym 4-[(4Z)-3-Chloro-1-[[4-[2-[(dimethyl-d6)amino]ethoxy]phenyl]phenylmethylene]propyl]phenol, 4-[(1Z)-3-chloro-1-[[4-[2-[(dimethyl-d6)amino]ethoxy]phenyl]phenylmethylene]propyl]phenol, EM 880
InChI Formula InChI=1S/C26H28ClNO2/c1-28(2)18-19-30-24-14-10-22(11-15-24)26(21-6-4-3-5-7-21)25(16-17-27)20-8-12-23(29)13-9-20/h3-15,29H,16-19H2,1-2H3/b26-25-/i1D3,2D3
SMILES [2H]C([2H])([2H])N(CCOc1ccc(cc1)\C(=C(\CCCl)/c2ccc(O)cc2)\c3ccccc3)C([2H])([2H])[2H]
IUPAC Name 4-[(Z)-1-[4-[2-[bis(trideuteriomethyl)amino]ethoxy]phenyl]-4-chloro-1-phenylbut-1-en-2-yl]phenol
Molecular Weight (g/mol) 427.996
Formula Weight 427.219
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.