missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Tolvaptan (Mixture of Diastereomers), TRC

Product Code. 30402171
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402171 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402171 Supplier Toronto Research Chemicals Supplier No. TRCH96989025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy Tolvaptan (Mixture of Diastereomers)
CAS 1587638-05-8
Molecular Formula C26 H25 Cl N2 O4
Quantity 25 mg
Synonym N-[4-[(7-Chloro-2,3,4,5-tetrahydro-4,5-dihydroxy-1H-1-benzazepin-1-yl)carbonyl]-3-methylphenyl]-2-methylbenzamide, 4-Hydroxy Tolvaptan (Mixture of Diastereomers)
InChI Formula InChI=1S/C26H25ClN2O4/c1-15-5-3-4-6-19(15)25(32)28-18-8-9-20(16(2)13-18)26(33)29-12-11-23(30)24(31)21-14-17(27)7-10-22(21)29/h3-10,13-14,23-24,30-31H,11-12H2,1-2H3,(H,28,32)
SMILES Cc1ccccc1C(=O)Nc2ccc(C(=O)N3CCC(O)C(O)c4cc(Cl)ccc34)c(C)c2
IUPAC Name N-[4-(7-chloro-4,5-dihydroxy-2,3,4,5-tetrahydro-1-benzazepine-1-carbonyl)-3-methylphenyl]-2-methylbenzamide
Molecular Weight (g/mol) 464.94
Formula Weight 464.1503
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.