missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Raloxifene, TRC

Product Code. 30393008
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393008 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393008 Supplier Toronto Research Chemicals Supplier No. TRCH95346010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy Raloxifene
CAS 185416-01-7
Molecular Formula C28 H27 N O4 S
Quantity 10 mg
Synonym [4-Hydroxy-2-(4-hydroxyphenyl)benzo[b]thien-3-yl][4-[2-(1-piperidinyl)ethoxy]phenyl]-methanone
InChI Formula InChI=1S/C28H27NO4S/c30-21-11-7-20(8-12-21)28-26(25-23(31)5-4-6-24(25)34-28)27(32)19-9-13-22(14-10-19)33-18-17-29-15-2-1-3-16-29/h4-14,30-31H,1-3,15-18H2
SMILES Oc1ccc(cc1)c2sc3cccc(O)c3c2C(=O)c4ccc(OCCN5CCCCC5)cc4
IUPAC Name [4-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(2-piperidin-1-ylethoxy)phenyl]methanone
Molecular Weight (g/mol) 473.583
Formula Weight 473.166
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.