missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Propranolol Beta-D-Glucuronide (Mixture of Diastereomers), TRC

Product Code. 30402588
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402588 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402588 Supplier Toronto Research Chemicals Supplier No. TRCH9525351MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 94731-13-2
Molecular Formula C22 H29 N O9
Quantity 1 mg
Synonym 4-Hydroxy Propranolol Glucuronide (Mixture of Diastereomers)
InChI Formula InChI=1S/C22H29NO9/c1-11(2)23-9-12(24)10-30-15-7-8-16(14-6-4-3-5-13(14)15)31-22-19(27)17(25)18(26)20(32-22)21(28)29/h3-8,11-12,17-20,22-27H,9-10H2,1-2H3,(H,28,29)/t12?,17-,18-,19+,20-,22+/m0/s1
SMILES CC(C)NCC(O)COc1ccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)c3ccccc13
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]naphthalen-1-yl]oxyoxane-2-carboxylic acid
Molecular Weight (g/mol) 451.47
Formula Weight 451.1842
Chemical Name or Material 4-Hydroxy Propranolol beta-D-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.