missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Propofol 1-O-β-D-Glucuronide Sodium Salt, TRC

Product Code. 30393682
Change view
Click to view available options
Quantity:
1 mg
2 mg
5 mg
Unit Size:
1mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393682 5 mg 5mg
30404946 2 mg 2mg
30392607 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393682 Supplier Toronto Research Chemicals Supplier No. TRCH9507785MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy Propofol 1-O-Beta-D-Glucuronide Sodium Salt
Analyte or Component Names 4-Hydroxy Propofol 1-O-Beta-D-Glucuronide Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C18 H25 Na O8
Synonym 4-Hydroxy-2,6-bis(1-methylethyl)phenol β-D-Glucopyranosiduronic Acid
InChI Formula InChI=1S/C18H26O8.Na/c1-7(2)10-5-9(19)6-11(8(3)4)15(10)25-18-14(22)12(20)13(21)16(26-18)17(23)24;/h5-8,12-14,16,18-22H,1-4H3,(H,23,24);/q;+1/p-1/t12-,13-,14+,16-,18+;/m0./s1
Solution Type Neat
SMILES CC(C)c1cc(O)cc(C(C)C)c1O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O[Na]
IUPAC Name [(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-hydroxy-2,6-di(propan-2-yl)phenoxy]oxane-2-carbonyl]oxysodium
Industry Type Pharmaceutical
Molecular Weight (g/mol) 392.376
Quantity 5 mg
Formula Weight 392.145
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.