missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Phenylbutazone, TRC

Product Code. 30389760
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389760 100 mg 100mg
30398310 5 mg -
30407356 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389760 Supplier Toronto Research Chemicals Supplier No. TRCH940060100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16860-43-8
Molecular Formula C19 H20 N2 O3
Molecular Weight (g/mol) 324.37
Synonym 4-Butyl-4-hydroxy-1,2-diphenylpyrazolidine-3,5-dione, Phenylbutazone Imp. B (EP)
IUPAC Name 4-butyl-4-hydroxy-1,2-diphenylpyrazolidine-3,5-dione
SMILES CCCCC1(O)C(=O)N(N(C1=O)c2ccccc2)c3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H20N2O3/c1-2-3-14-19(24)17(22)20(15-10-6-4-7-11-15)21(18(19)23)16-12-8-5-9-13-16/h4-13,24H,2-3,14H2,1H3
Formula Weight 324.1474
Chemical Name or Material 4-Butyl-4-hydroxy-1,2-diphenylpyrazolidine-3,5-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.