missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Omeprazole Sulfide, TRC

Product Code. 30135102
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135102 25 mg 25mg
30007237 5 mg 5mg
30011949 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135102 Supplier Toronto Research Chemicals Supplier No. TRCH94884525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103876-98-8
Molecular Formula C16H17N3O2S
Molecular Weight (g/mol) 315.39
Synonym 2-[[(6-Methoxy-1H-benzimidazol-2-yl)thio]methyl]-3,5-dimethyl-4-pyridinol, 2-[[(5-Methoxy-1H-benzimidazol-2-yl)thio]methyl]-3,5-dimethyl-4-pyridinol, Omeprazole Pyridone;
IUPAC Name 2-[(5-methoxy-1H-benzimidazol-2-yl)sulfanylmethyl]-3,5-dimethylpyridin-4-ol
SMILES COc1ccc2[nH]c(SCc3ncc(C)c(O)c3C)nc2c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H17N3O2S/c1-9-7-17-14(10(2)15(9)20)8-22-16-18-12-5-4-11(21-3)6-13(12)19-16/h4-7H,8H2,1-3H3,(H,17,20)(H,18,19)
Formula Weight 315.1041
Chemical Name or Material 4-Hydroxy Omeprazole Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.