missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-N-methylpiperidine, 98%

Product Code. 10010990 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10010990 500 g Glass bottle 500g
10051010 5 g Glass bottle 5g
10687462 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10010990 Supplier Thermo Scientific Acros Supplier No. 121515000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-52-5 | C6H13NO | 115.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-52-5
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
MDL Number MFCD00006500
InChI Key BAUWRHPMUVYFOD-UHFFFAOYSA-N
Synonym 4-hydroxy-1-methylpiperidine, n-methyl-4-piperidinol, 1-methyl-4-piperidinol, 4-hydroxy-n-methylpiperidine, n-methyl-4-hydroxypiperidine, 4-piperidinol, 1-methyl, 1-methyl-4-hydroxypiperidine, n-methylpiperidol, n-methyl-4-hydroxy piperidine, 4-hydroxy-1-methyl-piperidine
PubChem CID 66048
IUPAC Name 1-methylpiperidin-4-ol
SMILES CN1CCC(CC1)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29°C to 31°C
Color Brown to Colorless
Boiling Point 200°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4760 to 1.479
Quantity 500 g
Beilstein 21, I, 188
Solubility Information Solubility in water: miscible with water
Formula Weight 115.17
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 4-Hydroxy-N-methylpiperidine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hydroxy-N-methylpiperidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.