missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-N-isopropyl-N-methyltryptamine, TRC

Product Code. 30401567
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401567 5 mg 5mg
30395226 10 mg -
30403985 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401567 Supplier Toronto Research Chemicals Supplier No. TRCH9428805MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77872-43-6
Molecular Formula C14 H20 N2 O
Molecular Weight (g/mol) 232.32
Synonym 1H-Indol-4-ol, 3-[2-[methyl(1-methylethyl)amino]ethyl]-, 3-[2-[Methyl(1-methylethyl)amino]ethyl]-1H-indol-4-ol, 4-Hydroxy-N-isopropyl-N-methyltryptamine, 4-HO-MiPT, 4-Hydroxy-N-methyl-N-isopropyltryptamine
IUPAC Name 3-[2-[methyl(propan-2-yl)amino]ethyl]-1H-indol-4-ol
SMILES CC(C)N(C)CCc1c[nH]c2cccc(O)c12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H20N2O/c1-10(2)16(3)8-7-11-9-15-12-5-4-6-13(17)14(11)12/h4-6,9-10,15,17H,7-8H2,1-3H3
Formula Weight 232.1576
Chemical Name or Material 4-hydroxy MiPT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.