missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Fenofibric Acid, TRC

Product Code. 30399079
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399079 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399079 Supplier Toronto Research Chemicals Supplier No. TRCH94274510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy Fenofibric Acid
CAS 61002-27-5
Molecular Formula C17 H16 O5
Quantity 10 mg
Synonym 2-[4-(4-Hydroxybenzoyl)phenoxy]-2-methyl-propanoic Acid, LF 634
InChI Formula InChI=1S/C17H16O5/c1-17(2,16(20)21)22-14-9-5-12(6-10-14)15(19)11-3-7-13(18)8-4-11/h3-10,18H,1-2H3,(H,20,21)
SMILES CC(C)(Oc1ccc(cc1)C(=O)c2ccc(O)cc2)C(=O)O
IUPAC Name 2-[4-(4-hydroxybenzoyl)phenoxy]-2-methylpropanoic acid
Molecular Weight (g/mol) 300.306
Formula Weight 300.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.