missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Felbamate, TRC

Product Code. 30391069
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391069 100 mg 100mg
30394970 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391069 Supplier Toronto Research Chemicals Supplier No. TRCH942470100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109482-28-2
Molecular Formula C11H14N2O5
Molecular Weight (g/mol) 254.24
Synonym 2-(4-Hydroxyphenyl)-1,3-propanediyl Dicarbamate, W 2986
IUPAC Name [3-carbamoyloxy-2-(4-hydroxyphenyl)propyl] carbamate
SMILES NC(=O)OCC(COC(=O)N)c1ccc(O)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H14N2O5/c12-10(15)17-5-8(6-18-11(13)16)7-1-3-9(14)4-2-7/h1-4,8,14H,5-6H2,(H2,12,15)(H2,13,16)
Formula Weight 254.0903
Chemical Name or Material 4-Hydroxy Felbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.