missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Estradiol 1-N7-Guanine, TRC

Product Code. 30392878
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392878 10 mg
30391083 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392878 Supplier Toronto Research Chemicals Supplier No. TRCH94211510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178971-91-0
Molecular Formula C23H27N5O4
Molecular Weight (g/mol) 437.49
Synonym 2-Amino-1,7-dihydro-7-[(17β)-3,4,17-trihydroxyestra-1,3,5(10)-trien-1-yl]-6H-purin-6-one, 4-OH-E2-1-N7Gua
SMILES C[C@]12CC[C@H]3[C@H]([C@@H]1CC[C@@H]2O)CCC4=C3C(=CC(=C4O)O)N5C=NC6=C5C(=O)N=C(N6)N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H27N5O4/c1-23-7-6-11-10(13(23)4-5-16(23)30)2-3-12-17(11)14(8-15(29)19(12)31)28-9-25-20-18(28)21(32)27-22(24)26-20/h8-11,13,16,29-31H,2-7H2,1H3,(H3,24,26,27,32)/t10-,11+,13+,16+,23+/m1/s1
Formula Weight 437.2063
Chemical Name or Material 4-Hydroxy Estradiol 1-N7-Guanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.