missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Estradiol 1-N3-Adenine, TRC

Product Code. 30394014
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394014 1 mg
30393516 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394014 Supplier Toronto Research Chemicals Supplier No. TRCH9421001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 428506-88-1
Molecular Formula C23H27N5O3
Molecular Weight (g/mol) 421.49
Synonym (17β)-1-(6-Amino-3H-purin-3-yl)estra-1,3,5(10)-triene-3,4,17-triol, 4-OH-E2-1-N3Ade
IUPAC Name (8R,9S,13S,14S,17S)-1-(6-aminopurin-3-yl)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,4,17-triol
SMILES C[C@]12CC[C@H]3[C@H]([C@@H]1CC[C@@H]2O)CCC4=C3C(=CC(=C4O)O)N5C=NC(=C6C5=NC=N6)N

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C23H27N5O3/c1-23-7-6-12-11(14(23)4-5-17(23)30)2-3-13-18(12)15(8-16(29)20(13)31)28-10-27-21(24)19-22(28)26-9-25-19/h8-12,14,17,29-31H,2-7,24H2,1H3/t11-,12+,14+,17+,23+/m1/s1
Formula Weight 421.2114
Chemical Name or Material 4-Hydroxy Estradiol 1-N3-Adenine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.