missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Duloxetine, TRC

Product Code. 30011780
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011780 10 mg 10mg
30010851 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011780 Supplier Toronto Research Chemicals Supplier No. TRCH94180010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 662149-13-5
Molecular Formula C18 H19 N O2 S
Molecular Weight (g/mol) 313.41
Synonym 1-Naphthalenol, 4-[(1S)-3-(methylamino)-1-(2-thienyl)propoxy]-, 4-[(1S)-3-(Methylamino)-1-(2-thienyl)propoxy]-1-naphthalenol, (S)-N-Methyl-3-(4-hydroxynaphthalen-1-yloxy)-3-(thiophen-2-yl)propan-1-amine, 4-Hydroxy Duloxetine, 4-Hydroxyduloxetine, (S)-4-Hydroxyduloxetine
IUPAC Name 4-[(1S)-3-(methylamino)-1-thiophen-2-ylpropoxy]naphthalen-1-ol
SMILES CNCC[C@H](Oc1ccc(O)c2ccccc12)c3cccs3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H19NO2S/c1-19-11-10-17(18-7-4-12-22-18)21-16-9-8-15(20)13-5-2-3-6-14(13)16/h2-9,12,17,19-20H,10-11H2,1H3/t17-/m0/s1
Formula Weight 313.1136
Percent Purity >95%
Chemical Name or Material 4-Hydroxy Duloxetine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.