missing translation for 'onlineSavingsMsg'
Learn More

4’-Hydroxy Diclofenac, TRC

Product Code. 18094010
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18094010 1 mg 1mg
18003320 10 mg 10mg
18042010 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18094010 Supplier Toronto Research Chemicals Supplier No. TRCH8252251MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64118-84-9
Molecular Formula C14 H11 Cl2 N O3
Molecular Weight (g/mol) 312.15
Synonym Benzeneacetic acid, 2-[(2,6-dichloro-4-hydroxyphenyl)amino]-, 2-[(2,6-Dichloro-4-hydroxyphenyl)amino]benzeneacetic acid, 4'-Hydroxydiclofenac, Diclofenac-4-hydroxy, 4'-Hydroxy Diclofenac
IUPAC Name 2-[2-(2,6-dichloro-4-hydroxyanilino)phenyl]acetic acid
SMILES OC(=O)Cc1ccccc1Nc2c(Cl)cc(O)cc2Cl

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H11Cl2NO3/c15-10-6-9(18)7-11(16)14(10)17-12-4-2-1-3-8(12)5-13(19)20/h1-4,6-7,17-18H,5H2,(H,19,20)
Formula Weight 311.0116
Percent Purity >95%
Chemical Name or Material 4'-Hydroxydiclofenac
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.