missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Coumarin-d4, TRC

Product Code. 30240556
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240556 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240556 Supplier Toronto Research Chemicals Supplier No. TRCH92487225MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 106754-18-1
Molecular Formula C9H2D4O3
Quantity 25 mg
Synonym 4-Hydroxy-2H-1-benzopyran-2-one-5,6,7,8-d4, 4-Hydroxy-2H-1-benzopyran-2-one-d4, 4-Coumarinol-d4, 4-Hydroxy-2H-1-benzopyran-2-one-d4, 4-Hydroxy-2H-benzo[b]pyran-2-one-d4, 4-Hydroxy-2H-chromen-2-one-d4, 4-Hydroxychromen-2-one-d4, 4-Hydroxycoumariin-d4, 4-Hydroxycoumarin-d4, Benzotetronic Acid-d4, NSC 11889-d4
InChI Formula InChI=1S/C9H6O3/c10-7-5-9(11)12-8-4-2-1-3-6(7)8/h1-5,10H/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c([2H])c2C(=CC(=O)Oc2c1[2H])O
IUPAC Name 5,6,7,8-tetradeuterio-4-hydroxychromen-2-one
Molecular Weight (g/mol) 166.17
Formula Weight 166.0568
Chemical Name or Material 4-Hydroxy Coumarin-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.