missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy Clofentezine, TRC

Product Code. 30337616
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337616 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337616 Supplier Toronto Research Chemicals Supplier No. TRCC14824010MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Clofentezine-4-hydroxy
CAS 107573-61-5
Molecular Formula C14 H8 Cl2 N4 O
Quantity 10 mg
Synonym Phenol, 3-chloro-4-[6-(2-chlorophenyl)-1,2,4,5-tetrazin-3-yl], 1,2,4,5-Tetrazine, phenol deriv., 3-Chloro-4-[6-(2-chlorophenyl)-1,2,4,5-tetrazin-3-yl]phenol
InChI Formula InChI=1S/C14H8Cl2N4O/c15-11-4-2-1-3-9(11)13-17-19-14(20-18-13)10-6-5-8(21)7-12(10)16/h1-7,21H
SMILES Oc1ccc(c(Cl)c1)c2nnc(nn2)c3ccccc3Cl
IUPAC Name 3-chloro-4-[6-(2-chlorophenyl)-1,2,4,5-tetrazin-3-yl]phenol
Molecular Weight (g/mol) 319.15
Formula Weight 318.0075
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.