missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Carbazole, TRC

Product Code. 17963205
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17963205 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17963205 Supplier Toronto Research Chemicals Supplier No. TRCH8847251G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 52602-39-8
Molecular Formula C12H9NO
Quantity 1 g
Synonym 9H-Carbazol-4-ol, 4-Carbazolol (6CI), 4-Hydroxy-9H-carbazole, 4-Hydroxycarbazole, SKF 106023
InChI Formula InChI=1S/C12H9NO/c14-11-7-3-6-10-12(11)8-4-1-2-5-9(8)13-10/h1-7,13-14H
SMILES Oc1cccc2[nH]c3ccccc3c12
IUPAC Name 9H-carbazol-4-ol
Molecular Weight (g/mol) 183.21
Formula Weight 183.068
Chemical Name or Material 4-Hydroxycarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.