missing translation for 'onlineSavingsMsg'
Learn More

4’-Hydroxy Atomoxetine-d3, TRC

Product Code. 30394582
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394582 10 mg
30403199 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30394582 Supplier Toronto Research Chemicals Supplier No. TRCH80250210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217686-14-0
Molecular Formula C17H18D3NO2
Molecular Weight (g/mol) 274.37
Synonym 4-Hydroxyatomoxetine-d3, 3-Methyl-4-(3-methylamino-d3-1-phenyl-propoxy)-phenol
SMILES CC1=C(O[C@H](CCNC([2H])([2H])[2H])C2=CC=CC=C2)C=CC(O)=C1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H21NO2/c1-13-12-15(19)8-9-16(13)20-17(10-11-18-2)14-6-4-3-5-7-14/h3-9,12,17-19H,10-11H2,1-2H3/t17-/m1/s1/i2D3
Formula Weight 274.1761
Percent Purity >95%
Chemical Name or Material 4'-Hydroxy Atomoxetine-d3
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.