missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Alverine, TRC

Product Code. 30395913
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395913 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395913 Supplier Toronto Research Chemicals Supplier No. TRCH8279501MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy Alverine
CAS 142047-94-7
Molecular Formula C20H27NO
Quantity 1 mg
Synonym 4-[3-[Ethyl(3-phenylpropyl)amino]propyl]phenol
InChI Formula InChI=1S/C20H27NO/c1-2-21(16-6-10-18-8-4-3-5-9-18)17-7-11-19-12-14-20(22)15-13-19/h3-5,8-9,12-15,22H,2,6-7,10-11,16-17H2,1H3
SMILES CCN(CCCc1ccccc1)CCCc2ccc(O)cc2
IUPAC Name 4-[3-[ethyl(3-phenylpropyl)amino]propyl]phenol
Molecular Weight (g/mol) 297.43
Formula Weight 297.2093
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.