missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-7-methoxyquinazolin-6-yl Ester Acetic Acid, TRC

Product Code. 30402248
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402248 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402248 Supplier Toronto Research Chemicals Supplier No. TRCH825780500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy-7-methoxyquinazolin-6-yl ester acetic acid
CAS 179688-53-0
Molecular Formula C11 H10 N2 O4
Quantity 500 mg
Synonym 4(3H)-Quinazolinone, 6-(acetyloxy)-7-methoxy-, 4(1H)-Quinazolinone, 6-(acetyloxy)-7-methoxy- (9CI), 6-(Acetyloxy)-7-methoxy-4(3H)-quinazolinone, 3,4-Dihydro-4-oxo-6-acetoxy-7-methoxyquinazoline, 3,4-Dihydro-4-oxo-6-acetyloxy-7-methoxyquinazoline, 3,4-Dihydro-7-methoxy-4-oxoquinazolin-6-yl acetate, 6-Acetoxy-7-methoxy-3,4-dihydroquinazolin-4-one, 6-Acetoxy-7-methoxyquinazol-4-one, 6-Acetoxy-7-methoxyquinazolin-4(3H)-one, 6-Acetoxy-7-methoxyquinazolin-4-one, 7-Methoxy-4-oxo-3,4-dihydroquinazolin-6-yl acetate, Acetic acid 4-hydroxy-7-methoxyquinazolin-6-yl ester
InChI Formula InChI=1S/C11H10N2O4/c1-6(14)17-10-3-7-8(4-9(10)16-2)12-5-13-11(7)15/h3-5H,1-2H3,(H,12,13,15)
SMILES COc1cc2N=CNC(=O)c2cc1OC(=O)C
IUPAC Name (7-methoxy-4-oxo-3H-quinazolin-6-yl) acetate
Molecular Weight (g/mol) 234.21
Formula Weight 234.0641
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.