missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-7-methoxy-6-quinolinecarboxamide, TRC

Product Code. 30392793
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392793 100 mg
30395847 50 mg
30396537 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392793 Supplier Toronto Research Chemicals Supplier No. TRCH211040100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2110414-05-4
Molecular Formula C11 H10 N2 O3
Molecular Weight (g/mol) 218.209
Synonym 4-Hydroxy-7-Methoxyquinoline-6-Carboxamide
IUPAC Name 4-hydroxy-7-methoxyquinoline-6-carboxamide
SMILES COc1cc2nccc(O)c2cc1C(=O)N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H10N2O3/c1-16-10-5-8-6(4-7(10)11(12)15)9(14)2-3-13-8/h2-5H,1H3,(H2,12,15)(H,13,14)
Formula Weight 218.069
Chemical Name or Material 4-Hydroxy-7-methoxy-6-quinolinecarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.