missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylic acid, Tech., Thermo Scientific™

Product Code. 10222792
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10222792 1 g 1g
10282832 10 g 10g
10585544 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10222792 Supplier Thermo Scientific Maybridge Supplier No. KM09299DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175203-86-8
Molecular Formula C11H6F3NO4
Molecular Weight (g/mol) 273.167
MDL Number MFCD00219848
InChI Key GSUDZUNGTZIKPY-UHFFFAOYSA-N
Synonym 4-hydroxy-6-trifluoromethoxy quinoline-3-carboxylic acid, 4-oxo-6-trifluoromethoxy-1h-quinoline-3-carboxylic acid, 4-hydroxy-6-trifluoromethoxy-3-quinolinecarboxylic acid, 3-quinolinecarboxylic acid, 1,4-dihydro-4-oxo-6-trifluoromethoxy, maybridge1_005008, pubchem6093, buttpark 10\01-49, 4-hydroxy-6-trifluoromethoxy quinoline-3-, 4-hydroxy-6-trifluoromethoxy-3-quinolinecarboxylicacid, 3-quinolinecarboxylicacid, 4-hydroxy-6-trifluoromethoxy
PubChem CID 2775105
IUPAC Name 4-oxo-6-(trifluoromethoxy)-1H-quinoline-3-carboxylic acid
SMILES C1=CC2=C(C=C1OC(F)(F)F)C(=O)C(=CN2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 273.17
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-Hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.