missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methylpyran-2-one, TRC

Product Code. 30395738
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
30395738 500 g 500g
1 options
This item is not returnable. View return policy

Product Code. 30395738

Brand: Toronto Research Chemicals TRCH952270500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy-6-methylpyran-2-one
CAS 675-10-5
Molecular Formula C6H6O3
Quantity 500 g
Synonym 4-Hydroxy-6-methyl-2H-pyran-2-one, 3,5-Dihydroxy-sorbic acid Δ-lactone, 4-Hydroxy-6-methyl-2-pyrone, 4-Hydroxy-6-methyl-α-pyrone, 4-Hydroxy-6-methylpyran-2-one, 6-Methyl-4-hydroxy-2-pyrone, NSC 34625, Triacetic acid lactone
InChI Formula InChI=1S/C6H6O3/c1-4-2-5(7)3-6(8)9-4/h2-3,7H,1H3
SMILES CC1=CC(=CC(=O)O1)O
IUPAC Name 4-hydroxy-6-methylpyran-2-one
Molecular Weight (g/mol) 126.11
Formula Weight 126.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.