missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methylcoumarin, 98+%, Thermo Scientific Chemicals

Catalog Number p-3212094
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13252-83-0
C10H8O3
176.17
MFCD00085738
CSVFZDYJGBVNEX-UHFFFAOYSA-N
4-hydroxy-6-methylcoumarin, 4-hydroxy-6-methyl-2h-chromen-2-one, 6-methyl-4-hydroxycoumarin, 2h-1-benzopyran-2-one,4-hydroxy-6-methyl, 2h-1-benzopyran-2-one, 4-hydroxy-6-methyl, 2-hydroxy-6-methylchromen-4-one, 2-hydroxy-6-methyl-4h-chromen-4-one, 6-methyl-4-hydroxy coumarin, hydroxy-6-methylcoumarin,4
54686349
4-hydroxy-6-methylchromen-2-one
CC1=CC=C2OC(O)=CC(=O)C2=C1
This item is not returnable. View return policy
13252-83-0
C10H8O3
176.17
MFCD00085738
CSVFZDYJGBVNEX-UHFFFAOYSA-N
4-hydroxy-6-methylcoumarin, 4-hydroxy-6-methyl-2h-chromen-2-one, 6-methyl-4-hydroxycoumarin, 2h-1-benzopyran-2-one,4-hydroxy-6-methyl, 2h-1-benzopyran-2-one, 4-hydroxy-6-methyl, 2-hydroxy-6-methylchromen-4-one, 2-hydroxy-6-methyl-4h-chromen-4-one, 6-methyl-4-hydroxy coumarin, hydroxy-6-methylcoumarin,4
54686349
4-hydroxy-6-methylchromen-2-one
CC1=CC=C2OC(O)=CC(=O)C2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 261.0°C to 264.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: sparingly soluble in hot ethanol and methanol
Formula Weight 176.17
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 4-Hydroxy-6-methylcoumarin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'