missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methyl-3-nitro-2-pyridone, 97%

Product Code. 11408587
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408587 25 g 25g
11498577 5 g 5g
11418587 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408587 Supplier Thermo Scientific Alfa Aesar Supplier No. A19690.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4966-90-9 | C6H6N2O4 | 170.12 g/mol

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Solubility in N,N-DMF gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4966-90-9
Molecular Formula C6H6N2O4
Molecular Weight (g/mol) 170.12
MDL Number MFCD00167612
InChI Key QIKWTNPFTOEELW-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methyl-3-nitro-2-pyridone, 4-hydroxy-6-methyl-3-nitropyridin-2 1h-one, 6-methyl-3-nitropyridine-2,4-diol, 2,4-dihydroxy-6-methyl-3-nitropyridine, 2-hydroxy-6-methyl-3-nitro-1h-pyridin-4-one, 4-hydroxy-6-methyl-3-nitro-1,2-dihydropyridin-2-one, 4-hydroxy-6-methyl-3-nitro-2 1h-pyridinone, 4-hydroxy-6-methyl-3-nitrohydropyridin-2-one, pubchem3892, acmc-209khe
PubChem CID 54685619
IUPAC Name 4-hydroxy-6-methyl-3-nitro-1H-pyridin-2-one
SMILES CC1=CC(=O)C(=C(O)N1)[N+]([O-])=O

Specifications

Melting Point >250°C (decomposition)
Quantity 25 g
Beilstein 1529425
Formula Weight 170.13
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-6-methyl-3-nitro-2-pyridone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.