missing translation for 'onlineSavingsMsg'
Learn More

4-hydroxy-6-mercaptopyrazolo[3,4-d]pyrimidine, 98%, Thermo Scientific™

Product Code. 10705623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10705623 5 g Glass bottle 5g
10280653 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10705623 Supplier Thermo Scientific Acros Supplier No. 376630050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 24521-76-4
Molecular Formula C5H4N4OS
Molecular Weight (g/mol) 168.18
MDL Number MFCD00022782
InChI Key SXRSXYWROQWSGJ-UHFFFAOYSA-N
Synonym 4-hydroxy-6-mercaptopyrazolo 3,4-d pyrimidine, 6-mercapto-1h-pyrazolo 3,4-d pyrimidin-4-ol, 4-hydroxy-6-mercapto-1h-pyrazolo 3,4-d pyrimidine, 6-mercapto-4-hydroxy-1h-pyrazolo 3,4-d pyrimidine, 6-sulfanyl-1h,4h,5h-pyrazolo 3,4-d pyrimidin-4-one, 6-mercapto-1,5-dihydro-4h-pyrazolo 3,4-d pyrimidin-4-one, 1,5,6,7-tetrahydro-6-thioxo-4h-pyrazolo 3,4-d pyrimidin-4-one, 4h-pyrazolo 3,4-d pyrimidin-4-one, 1,5,6,7-tetrahydro-6-thioxo, 4h-pyrazolo 3,4-d pyrimidin-4-one,1,5,6,7-tetrahydro-6-thioxo
PubChem CID 3034349
IUPAC Name 6-sulfanylidene-1,2-dihydropyrazolo[3,4-d]pyrimidin-4-one
SMILES C1=C2C(=NC(=S)NC2=O)NN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >310.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 168.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-6-mercaptopyrazolo[3,4-d]pyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hydroxy-6-mercaptopyrazolo¢3,4-d!pyrimidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.