missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-5-methyl-3-furanone, TRC

Product Code. 30397881
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397881 5 g
30397316 100 g
30402241 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397881 Supplier Toronto Research Chemicals Supplier No. TRCH8252705G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19322-27-1
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
Synonym 4-Hydroxy-5-methyl-3(2H)-furanone, 4-Hydroxy-5-methyl-2,3-dihydrofuran-3-one, 5-Methyl-4-hydroxy-2,3-dihydrofuran-3-one, 5-Methyl-4-hydroxy-3(2H)-furanone, Norfuraneol, Norfuronol
IUPAC Name 4-hydroxy-5-methylfuran-3-one
SMILES CC1=C(O)C(=O)CO1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H6O3/c1-3-5(7)4(6)2-8-3/h7H,2H2,1H3
Formula Weight 114.0317
Chemical Name or Material 4-Hydroxy-5-methyl-3-furanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.