missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-5-methyl-1,3-benzenedisulfonic Acid Dipotassium Salt, TRC

Product Code. 30397052
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397052 500 mg 500mg
30394373 250 mg 250mg
30390098 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397052 Supplier Toronto Research Chemicals Supplier No. TRCH575740500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy-5-methyl-1,3-benzenedisulfonic Acid Dipotassium Salt
Analyte or Component Names 4-Hydroxy-5-methyl-1,3-benzenedisulfonic Acid Dipotassium Salt
Purity Grade Notes HPLC
Shipping Condition Room Temperature
Molecular Formula C7H6K2O7S2
Synonym potassium 4-hydroxy-5-methylbenzene-1,3-disulfonate
InChI Formula InChI=1S/C7H8O7S2.2K/c1-4-2-5(15(9,10)11)3-6(7(4)8)16(12,13)14;;/h2-3,8H,1H3,(H,9,10,11)(H,12,13,14);;/q;2*+1/p-2
Solution Type Neat
SMILES OC1=C(C)C=C(S(=O)(O[K])=O)C=C1S(=O)(O[K])=O
Industry Type Pharmaceutical
Molecular Weight (g/mol) 344.43
Quantity 500 mg
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.