missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-5-methyl-1,3-benzenedisulfonic Acid Dipotassium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxy-5-methyl-1,3-benzenedisulfonic Acid Dipotassium Salt |
| Analyte or Component Names | 4-Hydroxy-5-methyl-1,3-benzenedisulfonic Acid Dipotassium Salt |
| Purity Grade Notes | HPLC |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7H6K2O7S2 |
| Synonym | potassium 4-hydroxy-5-methylbenzene-1,3-disulfonate |
| InChI Formula | InChI=1S/C7H8O7S2.2K/c1-4-2-5(15(9,10)11)3-6(7(4)8)16(12,13)14;;/h2-3,8H,1H3,(H,9,10,11)(H,12,13,14);;/q;2*+1/p-2 |
| Solution Type | Neat |
| SMILES | OC1=C(C)C=C(S(=O)(O[K])=O)C=C1S(=O)(O[K])=O |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?