missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-5-iodopyrimidine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.
Solubility
Soluble in water (67 g/L) (25°C).
Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.
Solubility
Soluble in water (67 g/L) (25°C).
Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Molecular Formula | C11H12N2O3 |
| MDL Number | MFCD00064341 |
| Quantity | 250 mg |
| Sensitivity | Light sensitive |
| Synonym | 5-hydroxy-l-tryptophan, l-5-hydroxytryptophan, oxitriptan, 5-hydroxytryptophan, levothym, tript-oh, cincofarm, l-5-htp, pretonine, 2s-2-amino-3-5-hydroxy-1h-indol-3-yl propanoic acid |
| Solubility Information | Soluble in water (67g/L) (25°C). |
| InChI Key | LDCYZAJDBXYCGN-IMWMWJONNA-N |
| SMILES | N[C@@H](CC1=CNC2=CC=C(O)C=C12)C(O)=O |
| IUPAC Name | (2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid |
| Molecular Weight (g/mol) | 220.23 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction